This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285745_m1
See other 4932438A13RIK GE Assays ›
Gene Symbol:
4932438A13Rik
Gene Name:
RIKEN cDNA 4932438A13 gene
Gene Aliases:
4732443H21, B830039D19Rik, D630029K19Rik, FSA, Kiaa1109, Tweek
Chromosome Location:
Chr.3: 36863106 - 37053033 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285745_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4932438A13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172679.2 NP_766267.2 70 - 71 12382 105
GenBank mRNA AK052702.1 - 2 - 3 200 105
AK173100.1 - 14 - 15 2017 105
BC079623.1 - 11 - 12 1752 105

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene 4932438A13Rik

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
lipid metabolic process
spermatogenesis
lipid storage
fat cell differentiation
nucleus localization
adipose tissue development
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300001920.mm.1 NM_172679.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300001920.mm.1 NM_172679.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300001920.mm.1 TC0300000272.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300001921.mm.1 NM_172679.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300001921.mm.1 NM_172679.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300001921.mm.1 TC0300000272.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0