This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01285867_m1
See other RBBP5 GE Assays ›
Gene Symbol:
Rbbp5
Gene Name:
retinoblastoma binding protein 5
Gene Aliases:
4933411J24Rik, C330016J05
Chromosome Location:
Chr.1: 132477347 - 132505665 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01285867_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbbp5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172517.2 NP_766105.2 10 - 11 1234 129
GenBank mRNA AK049247.1 - 10 - 11 1221 129
AK154782.1 - 10 - 11 1235 129
AK170520.1 - 10 - 11 1232 129
AK171177.1 - 10 - 11 1239 129
BC030892.1 - 4 - 5 435 129
BC057632.1 - 11 - 12 1107 4918318 129
CV562303.1 - 6 - 7 667 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001195.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbbp5

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
chromatin modification
response to estrogen
histone H3-K4 methylation
protein binding
methylated histone binding
histone methyltransferase activity (H3-K4 specific)
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100009735.mm.1 NM_172517.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009735.mm.1 NM_172517.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100009735.mm.1 TC0100001195.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100009734.mm.1 NM_172517.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009734.mm.1 NM_172517.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100009734.mm.1 TC0100001195.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0