This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01286487_m1
See other TNFRSF11A GE Assays ›
Gene Symbol:
Tnfrsf11a
Gene Name:
tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Aliases:
Ly109, ODFR, OFE, Rank, TRANCE-R, mRANK
Chromosome Location:
Chr.1: 105780723 - 105847981 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01286487_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf11a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009399.3 NP_033425.3 7 - 8 762 90
GenBank mRNA AF019046.1 - 7 - 8 757 90
AK138990.1 - 7 - 8 768 90
BC019185.1 - 7 - 8 759 5025470 90

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf11a

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

ossification
adaptive immune response
multicellular organism development
programmed cell death
osteoclast differentiation
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
response to cytokine
response to tumor necrosis factor
positive regulation of JUN kinase activity
lymph node development
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
circadian temperature homeostasis
mammary gland alveolus development
response to interleukin-1
positive regulation of fever generation by positive regulation of prostaglandin secretion
TNFSF11-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade via TNFSF11-mediated signaling
transmembrane signaling receptor activity
tumor necrosis factor-activated receptor activity
protein binding
cytokine binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline