This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01287180_g1
See other PRSS56 GE Assays ›
Gene Symbol:
Prss56
Gene Name:
protease, serine 56
Gene Aliases:
1700027L20Rik
Chromosome Location:
Chr.1: 87183094 - 87188405 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01287180_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027084.1 NP_081360.1 12 - 13 1541 58
GenBank mRNA AK006434.1 - 4 - 5 427 58
JF323950.1 - 12 - 13 1541 58
JF698684.1 - 12 - 13 1541 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000808.mm.1

Set Membership:

Amplicon length less than or equal to 70 Protease 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
camera-type eye development
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P431543 AK006434.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P431543 AK006434.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90560 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P431543 AK006434.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P431543 AK006434.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P431539 AK006434.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P431543 AK006434.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P431543 AK006434.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133494 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108002 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00914674 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20416188 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20416201 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20416204 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00456822 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00188517 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20416187 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20416203 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20416208 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00428776 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00188524 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20416195 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00428775 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10348145 - -
Affymetrix HT MG-430 PM Array Plate 1429647_PM_at - -
Mouse Genome 430 2.0 Array 1429647_PM_at - -
Mouse Genome 430 2.0 Array 1429647_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69453_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69453_PM_at - -
Affymetrix Mouse Clariom S Array PSR0100006759.mm.1 NM_027084.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100006759.mm.1 TC0100000808.mm.1 MTA TC
Mouse Expression Set 430 1429647_PM_at - -
Mouse Expression Set 430 1429647_at - -
Affymetrix HT Mouse Genome 430B Array 1429647_at - -
Affymetrix HT Mouse Genome 430B Array 1429647_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 456526 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100510619 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2746177 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline