This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01287185_g1
See other PRSS56 GE Assays ›
Gene Symbol:
Prss56
Gene Name:
protease, serine 56
Gene Aliases:
1700027L20Rik
Chromosome Location:
Chr.1: 87183094 - 87188405 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01287185_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027084.1 NP_081360.1 7 - 8 864 79
GenBank mRNA JF323950.1 - 7 - 8 864 79
JF698684.1 - 7 - 8 864 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000808.mm.1

Set Membership:

Amplicon length between 71 and 85 Protease Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
camera-type eye development
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100006754.mm.1 NM_027084.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100006754.mm.1 TC0100000808.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-t8vjm:80/100.66.78.50:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline