This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01287448_g1
See other MGST3 GE Assays ›
Gene Symbol:
Mgst3
Gene Name:
microsomal glutathione S-transferase 3
Gene Aliases:
2010012L10Rik, 2010306B17Rik, 2700004G04Rik, AA516734, GST-III
Chromosome Location:
Chr.1: 167372384 - 167393797 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01287448_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mgst3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025569.1 NP_079845.1 3 - 4 248 88
GenBank mRNA AK003246.1 - 3 - 4 248 88
AK003309.1 - 3 - 4 234 88
AK003476.1 - 3 - 4 241 88
AK003492.1 - 3 - 4 241 88
AK008533.1 - 3 - 4 237 88
AK135742.1 - 3 - 4 223 88
BC029669.1 - 2 - 3 227 5372427 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mgst3

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
glutathione transferase activity
glutathione peroxidase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100028510.mm.1 NM_025569.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028510.mm.1 NM_025569.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028510.mm.1 TC0100003483.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100028511.mm.1 NM_025569.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028511.mm.1 NM_025569.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028511.mm.1 TC0100003483.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline