This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288197_m1
See other EIF2D GE Assays ›
Gene Symbol:
Eif2d
Gene Name:
eukaryotic translation initiation factor 2D
Gene Aliases:
D1Ertd5e, Lgtn
Chromosome Location:
Chr.1: 131153170 - 131187659 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288197_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136070.1 NP_001129542.1 4 - 5 589 55
NM_010709.3 NP_034839.2 4 - 5 589 55
GenBank mRNA AK034115.1 - 4 - 5 561 55
AK036154.1 - 4 - 5 585 55
AK046539.1 - 4 - 5 601 55
AK047005.1 - 4 - 5 584 55
AK078472.1 - 3 - 4 503 55
AK082748.1 - 4 - 5 604 55
AK083638.1 - 4 - 5 549 55
AK139052.1 - 4 - 5 589 55
AK161269.1 - 4 - 5 572 55
AK166085.1 - 4 - 5 601 55
AK167980.1 - 4 - 5 585 55
AK168120.1 - 4 - 5 555 55
AK170252.1 - 4 - 5 616 55
AK170448.1 - 4 - 5 601 55
AK171115.1 - 4 - 5 567 55
BC023709.1 - 4 - 5 595 55
BC025036.1 - 4 - 5 545 4935223 55
U58337.1 - 4 - 5 474 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001168.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2d

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
IRES-dependent translational initiation
translation
translational initiation
ribosome disassembly
RNA binding
translation initiation factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100009394.mm.1 NM_001136070.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009394.mm.1 NM_001136070.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009394.mm.1 NM_010709.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009394.mm.1 NM_010709.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100009394.mm.1 TC0100001168.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0