This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288207_m1
See other PIGR GE Assays ›
Gene Symbol:
Pigr
Gene Name:
polymeric immunoglobulin receptor
Gene Aliases:
-
Chromosome Location:
Chr.1: 130826684 - 130852249 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288207_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pigr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011082.3 NP_035212.2 1 - 2 120 72
GenBank mRNA AK128944.1 - 1 - 2 120 72
AK142656.1 - 1 - 2 122 72
BC013556.1 - 1 - 2 53 3601095 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK142656.1, BC013556.1, NM_011082.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pigr

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
immunoglobulin transcytosis in epithelial cells mediated by polymeric immunoglobulin receptor
epidermal growth factor receptor signaling pathway
Fc receptor signaling pathway
receptor clustering
polymeric immunoglobulin receptor activity
epidermal growth factor receptor binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 742602 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0