This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288210_g1
See other PIGR GE Assays ›
Gene Symbol:
Pigr
Gene Name:
polymeric immunoglobulin receptor
Gene Aliases:
-
Chromosome Location:
Chr.1: 130826684 - 130852249 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288210_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pigr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011082.3 NP_035212.2 5 - 6 1550 67
GenBank mRNA AK128944.1 - 5 - 6 1550 67
AK142656.1 - 5 - 6 1552 67
BC013556.1 - 5 - 6 1483 3601095 67
U06431.1 - 5 - 6 1454 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pigr

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
immunoglobulin transcytosis in epithelial cells mediated by polymeric immunoglobulin receptor
epidermal growth factor receptor signaling pathway
Fc receptor signaling pathway
receptor clustering
polymeric immunoglobulin receptor activity
epidermal growth factor receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0