This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288243_m1
See other FABP12 GE Assays ›
Gene Symbol:
Fabp12
Gene Name:
fatty acid binding protein 12
Gene Aliases:
1700008G05Rik
Chromosome Location:
Chr.3: 10244209 - 10301526 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288243_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fabp12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029310.1 NP_083586.1 3 - 4 396 105
GenBank mRNA AK005765.1 - 2 - 3 242 105
BC030910.1 - 3 - 4 396 4505673 105
EU733649.1 - 2 - 3 372 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001641.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fabp12

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
transporter activity
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300013049.mm.1 NM_029310.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013049.mm.1 NM_029310.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013049.mm.1 TC0300001641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300013050.mm.1 NM_029310.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013050.mm.1 NM_029310.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013050.mm.1 TC0300001641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300013055.mm.1 NM_029310.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013055.mm.1 NM_029310.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013055.mm.1 TC0300001641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300013054.mm.1 NM_029310.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013054.mm.1 NM_029310.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013054.mm.1 TC0300001641.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0