This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01288516_g1
See other GPX3 GE Assays ›
Gene Symbol:
Gpx3
Gene Name:
glutathione peroxidase 3
Gene Aliases:
AA960521, EGPx, GPx, GSHPx-3, GSHPx-P
Chromosome Location:
Chr.11: 54902854 - 54910382 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01288516_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpx3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008161.3 NP_032187.2 3 - 4 468 76
GenBank mRNA AK002219.1 - 3 - 4 397 76
AK002262.1 - 3 - 4 397 76
AK004942.1 - 3 - 4 397 76
AK146760.1 - 3 - 4 397 76
BC003339.1 - 3 - 4 380 3257333 76
BC037027.1 - 3 - 4 415 5370640 76
BC037134.1 - 3 - 4 394 76
BC049235.1 - 3 - 4 384 4221025 76
BC061950.1 - 3 - 4 372 6528998 76
CB601893.1 - 3 - 4 400 76
U13705.1 - 3 - 4 372 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000638.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpx3

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
response to oxidative stress
hydrogen peroxide catabolic process
protein homotetramerization
oxidation-reduction process
peroxidase activity
glutathione peroxidase activity
selenium binding
oxidoreductase activity
glutathione binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812595 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812597 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812598 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812596 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 U13705.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC037134.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC049235.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC061950.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC003339.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 U13705.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC037134.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC049235.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC061950.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292008 BC003339.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169572 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134607 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895039 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642717 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392325 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171405 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 U13705.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC037134.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC049235.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC061950.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC003339.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 U13705.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC037134.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC049235.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC061950.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292008 BC003339.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 U13705.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC037134.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC049235.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC061950.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC003339.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 U13705.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC037134.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC049235.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC061950.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292008 BC003339.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812602 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812576 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812577 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812605 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812598 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812573 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812591 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812606 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812586 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812595 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812589 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812615 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812569 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812603 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812609 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812593 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812604 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812607 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812596 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812610 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812614 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812600 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812579 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812572 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812584 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812588 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812583 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812590 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27812592 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171405 - -
Whole Mouse Genome G4122A A_51_P292008 U13705.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC037134.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC049235.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC061950.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC003339.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 U13705.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC037134.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC049235.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC061950.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P292008 BC003339.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111373 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171405 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09621286 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09621287 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09621282 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09621294 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09072113 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09017231 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812597 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812598 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812596 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27812595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812598 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812582 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812591 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812605 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812620 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812576 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812638 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27812579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812577 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812645 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09626810 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812632 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812620 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09072113 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812563 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812576 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09461128 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812603 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812611 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09240509 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812564 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09240523 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812643 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09621287 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812585 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812626 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812638 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812648 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09461129 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812624 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812607 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09621286 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812572 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09369968 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812566 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812640 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812654 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812573 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09240535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09254584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812598 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812610 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09662104 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09626809 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812565 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09618347 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812582 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812600 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09349934 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09017231 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812586 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812618 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812622 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812609 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09406656 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09621291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812604 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812646 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812581 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812614 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27812590 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10376203 - -
Affymetrix Mouse Gene 1.0 ST Array 10376206 - -
Affymetrix Mouse Gene 1.0 ST Array 10376205 - -
Affymetrix Mouse Gene 1.0 ST Array 10376204 - -
Affymetrix Mouse Gene 1.0 ST Array 10376207 - -
Affymetrix Mouse Gene 1.0 ST Array 10376202 - -
Affymetrix HT MG-430 PM Array Plate 1449106_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10376201 - -
Mouse Genome 430 2.0 Array 1449106_PM_at - -
Mouse Genome 430 2.0 Array 1449106_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14778_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14778_at - -
Affymetrix Mouse Clariom S Array PSR1100005166.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005166.mm.1 TC1100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005167.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005167.mm.1 TC1100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005163.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005163.mm.1 TC1100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005165.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005165.mm.1 TC1100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005161.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005161.mm.1 TC1100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005155.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005155.mm.1 TC1100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005160.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005160.mm.1 TC1100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100005164.mm.1 NM_008161.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100005164.mm.1 TC1100000638.mm.1 MTA TC
Mouse Expression Set 430 1449106_at - -
Mouse Expression Set 430 1449106_PM_at - -
Murine Genome U74v2 Set 101676_at - -
Murine Genome U74v2 Set 101676_PM_at - -
Murine Genome U74v2 Set 168706_PM_r_at - -
Murine Genome U74v2 Set 168706_r_at - -
Mouse Genome 430A 2.0 Array 1449106_PM_at - -
Mouse Genome 430A 2.0 Array 1449106_at - -
Affymetrix HT Mouse Genome 430A Array 1449106_at - -
Affymetrix HT Mouse Genome 430A Array 1449106_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 1340450 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001340450 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2715546 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2715546 - -
mouse-6 v1.1 (Illumina) scl41544.8_269-S - -
Illumina Sentrix mouse-6.v1 bead array 1340450 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl41544.8_269-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340450 - -
MouseRef-8 v2 Expression BeadChip ILMN_2715546 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2715546 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41544.8_269-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0