This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01289122_g1
See other YTHDF3 GE Assays ›
Gene Symbol:
Ythdf3
Gene Name:
YTH domain family 3
Gene Aliases:
9130022A11Rik
Chromosome Location:
Chr.3: 16183005 - 16217037 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01289122_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ythdf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145919.1 NP_001139391.1 1 - 2 334 144
NM_172677.3 NP_766265.3 1 - 2 334 144
NR_027375.1 1 - 2 334 293
GenBank mRNA AK039176.1 - 1 - 2 281 293
AK053736.1 - 1 - 2 253 293
AK137245.1 - 1 - 2 281 144
BC057158.1 - 1 - 2 245 5348717 293
BC067040.1 - 1 - 2 305 6853196 144
BC067042.1 - 1 - 2 267 6856248 144

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK039176.1, AK053736.1, AK137245.1, BC057158.1, BC067040.1, BC067042.1, NM_001145919.1, NM_172677.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ythdf3

Gene Ontology Categories:

Function(s) Process(es)

biological_process
RNA binding
poly(A) RNA binding
N6-methyladenosine-containing RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0