This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01289714_m1
See other PLCH1 GE Assays ›
Gene Symbol:
Plch1
Gene Name:
phospholipase C, eta 1
Gene Aliases:
BC042549, PLCeta1, Plcl3
Chromosome Location:
Chr.3: 63696232 - 63899677 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01289714_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plch1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177732.1 NP_001171203.1 5 - 6 725 154
NM_001177733.1 NP_001171204.1 5 - 6 725 154
NM_183191.3 NP_899014.2 5 - 6 725 154
GenBank mRNA AY691172.1 - 5 - 6 725 154
AY691173.1 - 5 - 6 725 154
AY691174.1 - 5 - 6 725 154
BC055005.1 - 5 - 6 802 6491217 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002071.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plch1

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
signal transduction
lipid catabolic process
intracellular signal transduction
phosphatidylinositol-mediated signaling
phosphatidylinositol phospholipase C activity
signal transducer activity
calcium ion binding
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
calcium-dependent phospholipase C activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300015644.mm.1 NM_001177732.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015644.mm.1 NM_001177732.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015644.mm.1 NM_001177733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015644.mm.1 NM_001177733.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015644.mm.1 NM_183191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015644.mm.1 NM_183191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015644.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015651.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015649.mm.1 NM_001177732.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015649.mm.1 NM_001177732.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015649.mm.1 NM_001177733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015649.mm.1 NM_001177733.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015649.mm.1 NM_183191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015649.mm.1 NM_183191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015649.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015650.mm.1 NM_001177732.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015650.mm.1 NM_001177732.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015650.mm.1 NM_001177733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015650.mm.1 NM_001177733.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015650.mm.1 NM_183191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015650.mm.1 NM_183191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015650.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015643.mm.1 NM_001177732.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015643.mm.1 NM_001177732.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015643.mm.1 NM_001177733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015643.mm.1 NM_001177733.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015643.mm.1 NM_183191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015643.mm.1 NM_183191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015643.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015645.mm.1 NM_001177732.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015645.mm.1 NM_001177732.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015645.mm.1 NM_001177733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015645.mm.1 NM_001177733.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015645.mm.1 NM_183191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015645.mm.1 NM_183191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015645.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015648.mm.1 NM_001177732.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015648.mm.1 NM_001177732.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015648.mm.1 NM_001177733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015648.mm.1 NM_001177733.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015648.mm.1 NM_183191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015648.mm.1 NM_183191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015648.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015653.mm.1 TC0300002071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015652.mm.1 NM_001177732.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015652.mm.1 NM_001177732.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015652.mm.1 NM_001177733.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015652.mm.1 NM_001177733.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015652.mm.1 NM_183191.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015652.mm.1 NM_183191.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015652.mm.1 TC0300002071.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0