This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01289728_m1
See other EIF2A GE Assays ›
Gene Symbol:
Eif2a
Gene Name:
eukaryotic translation initiation factor 2A
Gene Aliases:
D030048D22, D3Ertd194e
Chromosome Location:
Chr.3: 58525821 - 58557501 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01289728_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005509.2 NP_001005509.1 6 - 7 716 120
GenBank mRNA AK050971.1 - 6 - 7 433 120
AK078519.1 - 6 - 7 495 120
AK145275.1 - 6 - 7 672 120
AK161904.1 - 6 - 7 497 120
BC030447.1 - 5 - 6 385 5400637 120
BC082557.1 - 6 - 7 521 30606936 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000457.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2a

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translation
protein phosphorylation
positive regulation of signal transduction
SREBP signaling pathway
ribosome assembly
tRNA binding
translation initiation factor activity
ribosome binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300003160.mm.1 NM_001005509.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003160.mm.1 NM_001005509.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300003160.mm.1 TC0300000457.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0