This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290081_mH
See other SSBP2 GE Assays ›
Gene Symbol:
Ssbp2
Gene Name:
single-stranded DNA binding protein 2
Gene Aliases:
1500004K09Rik, 2310079I02Rik, 9330163K02Rik, A830008M03Rik, AU067692, AW558684, Hspc116, SSDP2
Chromosome Location:
Chr.13: 91460979 - 91706175 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290081_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024272.4 NP_077234.2 4 - 5 376 100
GenBank mRNA AK013177.1 - 4 - 5 302 100
AY037837.1 - 4 - 5 332 100
AY167988.1 - 4 - 5 286 100

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssbp2

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
DNA binding
single-stranded DNA binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006515.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006515.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006515.mm.1 TC1300001020.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0