This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290176_m1
See other CADM3 GE Assays ›
Gene Symbol:
Cadm3
Gene Name:
cell adhesion molecule 3
Gene Aliases:
BIgR, Igsf4b, Necl-1, Necl1, SynCAM3, Tsll1
Chromosome Location:
Chr.1: 173334254 - 173367695 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290176_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cadm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053199.3 NP_444429.1 3 - 4 580 89
GenBank mRNA AF195662.1 - 3 - 4 524 89
AK036352.1 - 4 - 5 636 89
AK038917.1 - 3 - 4 537 89
AK053077.1 - 3 - 4 534 89
AY059393.1 - 3 - 4 379 89
BC079837.1 - 4 - 5 620 89
BC115417.2 - 3 - 4 485 40093410 89
BC115418.2 - 3 - 4 485 40093417 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003587.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cadm3

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
protein localization
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100029429.mm.1 NM_053199.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029429.mm.1 NM_053199.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100029429.mm.1 TC0100003587.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029425.mm.1 NM_053199.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029425.mm.1 NM_053199.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100029425.mm.1 TC0100003587.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0