This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290188_g1
See other L3HYPDH GE Assays ›
Gene Symbol:
L3hypdh
Gene Name:
L-3-hydroxyproline dehydratase (trans-)
Gene Aliases:
2810055F11Rik
Chromosome Location:
Chr.12: 72071660 - 72085433 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290188_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: L3hypdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026038.2 NP_080314.1 3 - 4 959 101
GenBank mRNA AK018449.1 - 3 - 4 960 101
BC004753.1 - 3 - 4 1019 3584353 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001899.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene L3hypdh

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
lyase activity
hydro-lyase activity
proline racemase activity
trans-L-3-hydroxyproline dehydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200013025.mm.1 NM_026038.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013025.mm.1 NM_026038.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200013025.mm.1 TC1200001899.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0