This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290375_m1
See other NDUFA10 GE Assays ›
Gene Symbol:
Ndufa10
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10
Gene Aliases:
2900053E13Rik
Chromosome Location:
Chr.1: 92439708 - 92473758 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290375_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024197.1 NP_077159.1 5 - 6 690 92
GenBank mRNA AK005339.1 - 5 - 6 726 92
AK013675.1 - 5 - 6 733 92
AK046292.1 - 594 92
AK145979.1 - 5 - 6 746 92
BC003439.1 - 5 - 6 690 2647815 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002810.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufa10

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
nucleobase-containing compound metabolic process
transport
oxidation-reduction process
ATP binding
NADH dehydrogenase (ubiquinone) activity
nucleoside kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100022755.mm.1 NM_024197.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100022755.mm.1 NM_024197.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100022755.mm.1 TC0100002810.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0