This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290662_g1
See other EIF2D GE Assays ›
Gene Symbol:
Eif2d
Gene Name:
eukaryotic translation initiation factor 2D
Gene Aliases:
D1Ertd5e, Lgtn
Chromosome Location:
Chr.1: 131153170 - 131187659 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290662_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136070.1 NP_001129542.1 14 - 15 1803 81
GenBank mRNA AK030312.1 - 14 - 15 1814 81
AK046539.1 - 14 - 15 1815 81
AK083638.1 - 14 - 15 1763 81
AK139052.1 - 14 - 15 1803 81
AK161269.1 - 14 - 15 1786 81
AK166085.1 - 14 - 15 1816 81
AK167980.1 - 14 - 15 1799 81
AK168120.1 - 14 - 15 1769 81
AK170252.1 - 14 - 15 1830 81
AK170448.1 - 14 - 15 1815 81
AK171115.1 - 14 - 15 1781 81
BC023709.1 - 14 - 15 1756 81
BC024501.1 - 9 - 10 1117 81
BC025036.1 - 14 - 15 1759 4935223 81

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
IRES-dependent translational initiation
translation
translational initiation
ribosome disassembly
RNA binding
translation initiation factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0