This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01290768_g1
See other ARHGAP30 GE Assays ›
Gene Symbol:
Arhgap30
Gene Name:
Rho GTPase activating protein 30
Gene Aliases:
6030405P05Rik, Gm102, mFLJ00267
Chromosome Location:
Chr.1: 171388960 - 171410240 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01290768_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005508.2 NP_001005508.2 6 - 7 973 84
GenBank mRNA AK031319.1 - 6 - 7 744 84
AK147757.1 - 6 - 7 980 84
AK156351.1 - 6 - 7 962 84
AK220204.1 - 1 - 2 107 84
BC082573.1 - 6 - 7 921 30548035 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001590.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap30

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100013095.mm.1 NM_001005508.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100013095.mm.1 NM_001005508.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100013095.mm.1 TC0100001590.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0