This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291021_m1
See other SLCO4C1 GE Assays ›
Gene Symbol:
Slco4c1
Gene Name:
solute carrier organic anion transporter family, member 4C1
Gene Aliases:
9530051F04, C330017E21Rik, OATP-H, OATP-M1, OATP4C1, OATPX, PRO2176, SLC21A20, oatp-R
Chromosome Location:
Chr.1: 96818784 - 96872171 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291021_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco4c1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172658.3 NP_766246.1 9 - 10 1679 138
GenBank mRNA AK049253.1 - 9 - 10 1679 138
AK049944.1 - 1644 138
AY654588.1 - 9 - 10 1679 138
BC119022.1 - 9 - 10 1638 8733771 138

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002856.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco4c1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
multicellular organism development
spermatogenesis
organic anion transport
cell differentiation
anion transmembrane transport
transporter activity
organic anion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100023112.mm.1 NM_172658.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023112.mm.1 NM_172658.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100023112.mm.1 TC0100002856.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100023113.mm.1 NM_172658.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023113.mm.1 NM_172658.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100023113.mm.1 TC0100002856.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f9vrx:80/100.66.79.34:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0