This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291045_m1
See other IGSF8 GE Assays ›
Gene Symbol:
Igsf8
Gene Name:
immunoglobulin superfamily, member 8
Gene Aliases:
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384
Chromosome Location:
Chr.1: 172312407 - 172319841 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291045_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080419.1 NP_536344.1 2 - 3 562 135
GenBank mRNA AF411055.1 - 2 - 3 433 135
AF439263.1 - 2 - 3 545 135
AK140254.1 - 2 - 3 575 135
AK146104.1 - 2 - 3 575 135
AK146221.1 - 2 - 3 581 135
AK150239.1 - 2 - 3 581 135
AK158522.1 - 2 - 3 583 135
AK168363.1 - 2 - 3 581 135
AK183814.1 - 1 - 2 219 135
BC048387.1 - 2 - 3 562 6311441 135
BC060087.1 - 1190 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001610.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf8

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell motility
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100013386.mm.1 NM_080419.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100013386.mm.1 TC0100001610.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100013387.mm.1 NM_080419.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100013387.mm.1 TC0100001610.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline