This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291057_m1
See other GPR161 GE Assays ›
Gene Symbol:
Gpr161
Gene Name:
G protein-coupled receptor 161
Gene Aliases:
Gm208, Gm208Gpr, vl
Chromosome Location:
Chr.1: 165281174 - 165326746 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291057_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr161
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081126.1 NP_001074595.1 3 - 4 576 119
GenBank mRNA AK145635.1 - 2 - 3 600 119
AY255596.1 - 1 - 2 67 119
BC137659.1 - 2 - 3 514 8860678 119
EF197953.1 - 2 - 3 601 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001516.mm.1

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
multicellular organism development
sensory perception of chemical stimulus
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
positive regulation of cAMP biosynthetic process
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
signal transducer activity
G-protein coupled receptor activity
G-protein coupled amine receptor activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137794 EF197953.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137794 AK145635.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137794 BC137659.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P137794 NM_001081126.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244254 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760101 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203456 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355947 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781322 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55287 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802437 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900046 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410926 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255002 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959556 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651708 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129228 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215579 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184268 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82232 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72227 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 169373 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12068 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113281 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137794 EF197953.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137794 AK145635.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137794 BC137659.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P137794 NM_001081126.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04339986 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113281 - -
Whole Mouse Genome G4122A A_52_P375503 - -
Whole Mouse Genome G4122A A_52_P243627 EF197953.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243627 AK145635.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243627 AY255596.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243627 BC137659.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243627 NM_001081126.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793792 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793982 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793991 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793966 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793814 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793983 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793933 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793907 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793952 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793838 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793794 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793960 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793801 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793854 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793880 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794003 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793868 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793951 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794016 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793992 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793975 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793821 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793971 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793828 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794011 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793784 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793785 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793877 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793826 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793819 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793980 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793856 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793989 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793789 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793773 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793786 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793808 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793964 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793937 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793950 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793962 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793976 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20793864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20794008 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00910380 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793839 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793969 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793826 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793761 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793989 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00122543 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793815 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00910397 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793829 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793794 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00332190 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793814 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20794004 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793759 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793779 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00888570 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00479400 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793823 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01134727 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00571080 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00033795 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00910387 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793819 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793987 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793771 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793833 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793790 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793828 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01134725 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793963 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00144422 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793765 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793831 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20794003 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793832 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00033789 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793760 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793956 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00571075 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793841 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793961 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00566415 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00910385 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793952 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00479401 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793966 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793774 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793960 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20794008 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793768 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00457937 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793781 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793792 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00751630 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793824 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793801 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793800 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793951 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00122544 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00059045 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00920499 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00017184 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793818 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793803 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00033790 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793958 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793769 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793954 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793798 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20793964 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00144421 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00571074 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00144420 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00539254 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00472487 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10351299 - -
Affymetrix Mouse Gene 1.0 ST Array 10351302 - -
Affymetrix Mouse Gene 1.0 ST Array 10351301 - -
Affymetrix Mouse Gene 1.0 ST Array 10351303 - -
Affymetrix Mouse Gene 1.0 ST Array 10351300 - -
Affymetrix HT MG-430 PM Array Plate 1447353_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10351298 - -
Mouse Genome 430 2.0 Array 1447353_at - -
Mouse Genome 430 2.0 Array 1447353_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 240888_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 240888_at - -
Affymetrix Mouse Clariom S Array PSR0100012483.mm.1 NM_001081126.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100012483.mm.1 TC0100001516.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100012477.mm.1 NM_001081126.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100012477.mm.1 TC0100001516.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100012479.mm.1 NM_001081126.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100012479.mm.1 TC0100001516.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100012482.mm.1 NM_001081126.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100012482.mm.1 TC0100001516.mm.1 MTA TC
Mouse Expression Set 430 1447353_PM_at - -
Mouse Expression Set 430 1447353_at - -
Affymetrix HT Mouse Genome 430B Array 1447353_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1447353_at - -
Array Name Probe IDs Targets Target Type
MouseRef-8 v2 Expression BeadChip ILMN_3096982 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3096982 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0