This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291339_m1
See other LGR6 GE Assays ›
Gene Symbol:
Lgr6
Gene Name:
leucine-rich repeat-containing G protein-coupled receptor 6
Gene Aliases:
A530037C04Rik, D830026M09
Chromosome Location:
Chr.1: 134983300 - 135105374 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291339_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lgr6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033409.3 NP_001028581.1 7 - 8 924 95
GenBank mRNA AK085901.1 - 7 - 8 882 95
AK137380.1 - 7 - 8 925 95
BC141210.1 - 7 - 8 876 9055822 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003119.mm.1

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
Wnt signaling pathway
positive regulation of Wnt signaling pathway
positive regulation of cell migration
molecular_function
signal transducer activity
G-protein coupled receptor activity
protein-hormone receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100025152.mm.1 NM_001033409.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025152.mm.1 NM_001033409.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100025152.mm.1 TC0100003119.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0