This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291350_m1
See other PTPN7 GE Assays ›
Gene Symbol:
Ptpn7
Gene Name:
protein tyrosine phosphatase, non-receptor type 7
Gene Aliases:
BPTP-4, C920001D21Rik, HEPTP, LC-PTP, LPTP
Chromosome Location:
Chr.1: 135131840 - 135145328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291350_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177081.3 NP_796055.1 7 - 8 832 61
GenBank mRNA AK041415.1 - 4 - 5 2873 61
AK049175.1 - 6 - 7 683 61
AK083305.1 - 7 - 8 832 61
BC098475.1 - 7 - 8 886 30044369 61
BC138194.1 - 6 - 7 768 8861213 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001244.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn7

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
peptidyl-tyrosine dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
phosphatase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 BC098475.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 BC138194.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 AK049175.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 AK083305.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 NM_177081.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 BC098475.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 BC138194.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 AK049175.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 AK083305.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P332487 NM_177081.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167360 AK083305.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167360 AK083305.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92849 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902454 NM_146993.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902454 NM_146996.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260652 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270828 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951442 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711776 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830503 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118903 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151309 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129145 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167360 AK083305.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167360 AK083305.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 BC098475.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 BC138194.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 AK049175.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 AK083305.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 NM_177081.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 BC098475.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 BC138194.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 AK049175.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 AK083305.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P332487 NM_177081.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167360 AK083305.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167360 AK083305.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637673 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637747 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637686 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637730 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637682 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637680 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637694 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637745 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637752 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637753 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637683 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637674 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637676 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637735 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637741 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637733 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637738 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637679 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637746 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637743 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637734 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637687 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637692 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637736 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637693 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637690 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637678 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637750 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637755 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637688 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637684 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20637677 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151309 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129145 - -
Whole Mouse Genome G4122A A_51_P167360 AK083305.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P167360 AK083305.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 BC098475.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 BC138194.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 AK049175.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 AK083305.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 NM_177081.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 BC098475.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 BC138194.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 AK049175.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 AK083305.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P332487 NM_177081.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109393 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151309 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129145 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00770178 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00770160 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00770182 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00770155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637788 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637743 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637741 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637776 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637823 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637799 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637811 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637790 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637784 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637773 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637824 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637812 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637735 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637734 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637801 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637725 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637763 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637682 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637786 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637807 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637821 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637745 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637690 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20637809 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637697 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637694 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637727 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00696874 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00464913 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770187 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00626498 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637705 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00403733 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637702 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00464912 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637680 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770178 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637729 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637716 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637730 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637748 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637699 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637700 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00790042 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637693 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770155 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770176 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00696875 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637725 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637735 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00626499 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637733 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770173 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637715 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770182 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00023013 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00403735 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00202876 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770160 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770177 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637709 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637723 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637734 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00403732 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637743 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637736 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00770180 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637750 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00696878 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637696 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637754 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637752 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00626501 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00464910 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00555882 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00696880 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20637714 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10350106 - -
Affymetrix Mouse Gene 1.0 ST Array 10350103 - -
Affymetrix Mouse Gene 1.0 ST Array 10350104 - -
Affymetrix Mouse Gene 1.0 ST Array 10350107 - -
Affymetrix Mouse Gene 1.0 ST Array 10350110 - -
Affymetrix Mouse Gene 1.0 ST Array 10350108 - -
Affymetrix Mouse Gene 1.0 ST Array 10350112 - -
Affymetrix Mouse Gene 1.0 ST Array 10350109 - -
Affymetrix Mouse Gene 1.0 ST Array 10350105 - -
Affymetrix Mouse Gene 1.0 ST Array 10350111 - -
Affymetrix Mouse Gene 1.1 ST Array 10350102 - -
Affymetrix Mouse Clariom S Array PSR0100010228.mm.1 NM_177081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010228.mm.1 NM_177081.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010228.mm.1 TC0100001244.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100010232.mm.1 NM_177081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010232.mm.1 NM_177081.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010232.mm.1 TC0100001244.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100010230.mm.1 NM_177081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010230.mm.1 NM_177081.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010230.mm.1 TC0100001244.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100010229.mm.1 NM_177081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010229.mm.1 NM_177081.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010229.mm.1 TC0100001244.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100010227.mm.1 NM_177081.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010227.mm.1 NM_177081.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010227.mm.1 TC0100001244.mm.1 MTA TC
Murine Genome U74v2 Set 135612_PM_at - -
Murine Genome U74v2 Set 135612_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 300305 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102480040 - -
Illumina mouse-6 v1.0 expression beadchip 3450110 - -
Illumina mouse-6 v1.0 expression beadchip 102480040 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003450110 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235056 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2455657 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1235056 - -
mouse-6 v1.1 (Illumina) scl000948.1_167-S - -
mouse-6 v1.1 (Illumina) scl0320139.8_83-S - -
Illumina Sentrix mouse-6.v1 bead array 3450110 - -
Illumina Sentrix mouse-6.v1 bead array 102480040 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0320139.8_83-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102480040 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450110 - -
MouseRef-8 v2 Expression BeadChip ILMN_1235056 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235056 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0