This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291495_m1
See other VAMP4 GE Assays ›
Gene Symbol:
Vamp4
Gene Name:
vesicle-associated membrane protein 4
Gene Aliases:
D1Ertd147e
Chromosome Location:
Chr.1: 162570404 - 162599086 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291495_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vamp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016796.3 NP_058076.2 2 - 3 167 91
GenBank mRNA AK002977.1 - 2 - 3 169 91
AK011678.1 - 2 - 3 147 91
AK018344.1 - 2 - 3 178 91
AK031151.1 - 2 - 3 169 91
BC138753.1 - 1 - 2 122 8861775 91
BC138754.1 - 1 - 2 122 8861776 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001479.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vamp4

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
exocytosis
vesicle fusion
vesicle-mediated transport
SNARE complex assembly
regulation of Golgi to plasma membrane protein transport
Golgi to plasma membrane protein transport
Golgi ribbon formation
toxin transport
SNARE binding
SNAP receptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100012164.mm.1 NM_016796.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100012164.mm.1 NM_016796.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100012164.mm.1 TC0100001479.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 930302 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0