This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291684_m1
See other ACMSD GE Assays ›
Gene Symbol:
Acmsd
Gene Name:
amino carboxymuconate semialdehyde decarboxylase
Gene Aliases:
-
Chromosome Location:
Chr.1: 127729413 - 127767564 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291684_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acmsd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033041.2 NP_001028213.1 5 - 6 564 72
GenBank mRNA AB071419.1 - 5 - 6 491 72
AK143962.1 - 5 - 6 566 72
BC139281.1 - 5 - 6 515 8862303 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acmsd

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
quinolinate metabolic process
aminocarboxymuconate-semialdehyde decarboxylase activity
catalytic activity
lyase activity
carboxy-lyase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0