This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291687_m1
See other ACMSD GE Assays ›
Gene Symbol:
Acmsd
Gene Name:
amino carboxymuconate semialdehyde decarboxylase
Gene Aliases:
-
Chromosome Location:
Chr.1: 127729413 - 127767564 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291687_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acmsd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033041.2 NP_001028213.1 8 - 9 928 89
GenBank mRNA AK143962.1 - 8 - 9 930 89
AK165228.1 - 2 - 3 294 89
BC139281.1 - 8 - 9 879 8862303 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001123.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acmsd

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
quinolinate metabolic process
aminocarboxymuconate-semialdehyde decarboxylase activity
catalytic activity
lyase activity
carboxy-lyase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100009086.mm.1 NM_001033041.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009086.mm.1 NM_001033041.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100009086.mm.1 TC0100001123.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0