This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291688_g1
See other ACMSD GE Assays ›
Gene Symbol:
Acmsd
Gene Name:
amino carboxymuconate semialdehyde decarboxylase
Gene Aliases:
-
Chromosome Location:
Chr.1: 127729413 - 127767564 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291688_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acmsd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033041.2 NP_001028213.1 9 - 10 1015 147
GenBank mRNA AK143962.1 - 9 - 10 1017 147
AK165228.1 - 3 - 4 381 147
BC139281.1 - 9 - 10 966 8862303 147

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001123.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acmsd

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
quinolinate metabolic process
aminocarboxymuconate-semialdehyde decarboxylase activity
catalytic activity
lyase activity
carboxy-lyase activity
metal ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161347 AK165228.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161347 NM_001033041.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2161347 AK143962.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725032 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881224 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888642 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354764 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969218 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242781 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865698 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469135 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684737 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344943 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867321 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962886 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852339 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541265 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397052 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503344 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432135 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638125 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417084 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912010 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343300 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946093 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123709 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63789 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99591 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126544 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164615 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116224 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161347 AK165228.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161347 NM_001033041.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2161347 AK143962.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596665 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596636 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596651 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596647 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596638 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596661 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596663 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596645 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596650 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596656 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596657 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596658 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596664 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596629 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596640 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596634 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596643 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596631 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596648 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596642 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596633 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596654 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20596632 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164615 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04255864 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00180283 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00180304 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04255881 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04255943 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116224 - -
Whole Mouse Genome G4122A A_51_P399873 BC139281.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P399873 NM_001033041.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P399873 AB071419.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P399873 AK143962.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116224 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596704 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596778 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596896 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596854 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596725 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596792 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596742 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596894 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596689 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596787 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596734 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596868 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596794 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596681 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596740 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596786 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596788 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596793 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596747 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596817 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596776 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596790 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596844 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596898 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596731 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596784 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596819 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596874 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596838 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596723 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596842 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596744 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596877 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596821 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20596661 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00618926 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00040858 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596654 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01044030 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596735 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596616 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596683 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596734 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596640 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00525957 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596650 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596656 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596613 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00380030 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00178109 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596668 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00618916 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01014439 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596822 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00249475 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596647 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596669 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00618920 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596681 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00341914 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00420665 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596820 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596679 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596832 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00984842 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00249477 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596648 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596748 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596659 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596674 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596667 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596672 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596646 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01044029 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00984836 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596634 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596644 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596657 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596817 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596633 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596830 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596675 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00178110 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00341910 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596643 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596649 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00144882 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00618921 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596742 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596813 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596661 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596736 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00911566 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00283413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00618927 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00420662 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00420664 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00382007 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596825 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01145267 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596658 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596835 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00984838 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596610 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596671 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00249470 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596833 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596609 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20596831 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10349432 - -
Affymetrix Mouse Gene 1.0 ST Array 10349433 - -
Affymetrix Mouse Gene 1.0 ST Array 10349439 - -
Affymetrix Mouse Gene 1.0 ST Array 10349436 - -
Affymetrix Mouse Gene 1.0 ST Array 10349441 - -
Affymetrix Mouse Gene 1.0 ST Array 10349437 - -
Affymetrix Mouse Gene 1.0 ST Array 10349440 - -
Affymetrix Mouse Gene 1.0 ST Array 10349434 - -
Affymetrix Mouse Gene 1.0 ST Array 10349438 - -
Affymetrix Mouse Gene 1.0 ST Array 10349435 - -
Affymetrix Mouse Gene 1.1 ST Array 10349431 - -
Affymetrix Mouse Clariom S Array PSR0100009088.mm.1 NM_001033041.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009088.mm.1 NM_001033041.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100009088.mm.1 TC0100001123.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100009087.mm.1 NM_001033041.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009087.mm.1 NM_001033041.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100009087.mm.1 TC0100001123.mm.1 MTA TC
Murine Genome U74v2 Set 132132_at - -
Murine Genome U74v2 Set 132132_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 834496 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1450538 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2626393 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2626393 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0