This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291819_m1
See other AGR3 GE Assays ›
Gene Symbol:
Agr3
Gene Name:
anterior gradient 3
Gene Aliases:
BCMP11, E030025L21Rik, Gm888
Chromosome Location:
Chr.12: 35925621 - 35949737 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291819_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207531.3 NP_997414.2 1 - 2 108 95
GenBank mRNA BE650904.1 - 1 - 2 108 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_207531.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Agr3

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
dystroglycan binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200002361.mm.1 NM_207531.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002361.mm.1 NM_207531.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002361.mm.1 TC1200000314.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002359.mm.1 NM_207531.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002359.mm.1 NM_207531.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002359.mm.1 TC1200000314.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0