This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01291977_m1
See other STK25 GE Assays ›
Gene Symbol:
Stk25
Gene Name:
serine/threonine kinase 25 (yeast)
Gene Aliases:
1500019J11Rik, AU018434, C86992, Ste20-like, Ysk1
Chromosome Location:
Chr.1: 93620751 - 93654216 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01291977_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stk25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021537.3 NP_067512.3 10 - 11 1335 74
GenBank mRNA AF004934.1 - 9 - 10 1173 74
AK148041.1 - 10 - 11 1309 74
AK152917.1 - 10 - 11 1278 74
BC029875.1 - 9 - 10 2053 74
BC052913.1 - 10 - 11 1270 30057101 74
BC071218.1 - 10 - 11 1330 4012695 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002836.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stk25

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
establishment or maintenance of cell polarity
regulation of mitotic cell cycle
phosphorylation
signal transduction by protein phosphorylation
stress-activated protein kinase signaling cascade
activation of protein kinase activity
positive regulation of stress-activated MAPK cascade
intrinsic apoptotic signaling pathway in response to hydrogen peroxide
response to hydrogen peroxide
regulation of apoptotic process
protein autophosphorylation
positive regulation of axonogenesis
Golgi localization
establishment of Golgi localization
Golgi reassembly
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100023013.mm.1 NM_021537.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100023013.mm.1 NM_021537.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100023013.mm.1 TC0100002836.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0