This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292365_m1
See other NPL GE Assays ›
Gene Symbol:
Npl
Gene Name:
N-acetylneuraminate pyruvate lyase
Gene Aliases:
0610033B02Rik
Chromosome Location:
Chr.1: 153503016 - 153549714 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292365_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028749.1 NP_083025.1 9 - 10 728 125
GenBank mRNA AK002734.1 - 9 - 10 728 125
BC022734.1 - 9 - 10 724 4506776 125

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Npl

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
metabolic process
N-acetylneuraminate catabolic process
catalytic activity
N-acetylneuraminate lyase activity
lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0