This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292950_m1
See other SEC23A GE Assays ›
Gene Symbol:
Sec23a
Gene Name:
SEC23A (S. cerevisiae)
Gene Aliases:
Msec23, Sec23r
Chromosome Location:
Chr.12: 58958383 - 59012026 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292950_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec23a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009147.2 NP_033173.2 3 - 4 472 81
GenBank mRNA AK028152.1 - 3 - 4 360 81
AY082671.1 - 3 - 4 346 81
BC034610.1 - 3 - 4 472 5364796 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001813.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec23a

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
ER to Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
protein binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200012415.mm.1 NM_009147.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200012415.mm.1 NM_009147.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200012415.mm.1 TC1200001813.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0