This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292993_m1
See other DLD GE Assays ›
Gene Symbol:
Dld
Gene Name:
dihydrolipoamide dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.12: 31331562 - 31351471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292993_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007861.5 NP_031887.2 1 - 2 169 70
GenBank mRNA AK014092.1 - 1 - 2 112 70
AK117104.1 - 1 - 2 113 70
AK135807.1 - 1 - 2 112 70
AK136193.1 - 1 - 2 113 70
AK153399.1 - 1 - 2 113 70
AK167885.1 - 1 - 2 113 70
AK168157.1 - 1 - 2 134 70
AK168875.1 - 1 - 2 54 70
BC003368.1 - 1 - 2 96 3589344 70
BY251665.1 - 1 - 2 136 70
BY282701.1 - 1 - 2 172 70
U73445.1 - 1 - 2 146 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001593.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dld

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

acetyl-CoA biosynthetic process from pyruvate
2-oxoglutarate metabolic process
mitochondrial electron transport, NADH to ubiquinone
proteolysis
gastrulation
lipoate metabolic process
regulation of membrane potential
cell redox homeostasis
sperm capacitation
dihydrolipoamide metabolic process
oxidation-reduction process
dihydrolipoyl dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor
lipoamide binding
flavin adenine dinucleotide binding
NAD binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200011038.mm.1 NM_007861.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200011038.mm.1 TC1200001593.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011037.mm.1 NM_007861.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200011037.mm.1 TC1200001593.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011039.mm.1 NM_007861.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200011039.mm.1 TC1200001593.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011040.mm.1 NM_007861.5 Refseq
Affymetrix Mouse Clariom S Array PSR1200011040.mm.1 TC1200001593.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 788379 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8t6tc:80/100.66.75.151:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0