This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01292996_g1
See other DLD GE Assays ›
Gene Symbol:
Dld
Gene Name:
dihydrolipoamide dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.12: 31331562 - 31351471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01292996_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dld
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007861.5 NP_031887.2 4 - 5 390 76
GenBank mRNA AK117104.1 - 4 - 5 334 76
AK135807.1 - 4 - 5 333 76
AK136193.1 - 4 - 5 334 76
AK153399.1 - 4 - 5 334 76
AK167885.1 - 4 - 5 334 76
AK168157.1 - 4 - 5 355 76
U73445.1 - 4 - 5 367 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dld

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

acetyl-CoA biosynthetic process from pyruvate
2-oxoglutarate metabolic process
mitochondrial electron transport, NADH to ubiquinone
proteolysis
gastrulation
lipoate metabolic process
regulation of membrane potential
cell redox homeostasis
sperm capacitation
dihydrolipoamide metabolic process
oxidation-reduction process
dihydrolipoyl dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, NAD(P) as acceptor
lipoamide binding
flavin adenine dinucleotide binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0