This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01293548_m1
See other CDHR3 GE Assays ›
Gene Symbol:
Cdhr3
Gene Name:
cadherin-related family member 3
Gene Aliases:
1110049B09Rik, BB172694, Cdh28
Chromosome Location:
Chr.12: 33033796 - 33093037 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01293548_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdhr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024478.1 NP_001019649.1 5 - 6 712 74
GenBank mRNA AK047666.1 - 5 - 6 713 74
AK049806.1 - 5 - 6 714 74
BC138911.1 - 5 - 6 640 8861933 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001615.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdhr3

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
biological_process
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200011260.mm.1 NM_001024478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011260.mm.1 NM_001024478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011260.mm.1 TC1200001615.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011258.mm.1 NM_001024478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011258.mm.1 NM_001024478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011258.mm.1 TC1200001615.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011261.mm.1 NM_001024478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011261.mm.1 NM_001024478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011261.mm.1 TC1200001615.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011259.mm.1 NM_001024478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011259.mm.1 NM_001024478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011259.mm.1 TC1200001615.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011263.mm.1 NM_001024478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011263.mm.1 NM_001024478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011263.mm.1 TC1200001615.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011262.mm.1 NM_001024478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011262.mm.1 NM_001024478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011262.mm.1 TC1200001615.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011257.mm.1 NM_001024478.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011257.mm.1 NM_001024478.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011257.mm.1 TC1200001615.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline