This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01294640_m1
See other PHYKPL GE Assays ›
Gene Symbol:
Phykpl
Gene Name:
5-phosphohydroxy-L-lysine phospholyase
Gene Aliases:
2900006B13Rik, Agxt2l2
Chromosome Location:
Chr.11: 51583983 - 51603269 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01294640_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phykpl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028398.2 NP_082674.1 6 - 7 685 65
GenBank mRNA AK013489.1 - 6 - 7 639 65
AK085984.1 - 5 - 6 711 65
AK133235.1 - 632 65
AK160557.1 - 6 - 7 654 65
BC024461.1 - 6 - 7 639 4975449 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000571.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phykpl

Gene Ontology Categories:

Function(s) Process(es)

biological_process
catalytic activity
transaminase activity
transferase activity
lyase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100004444.mm.1 NM_028398.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004444.mm.1 NM_028398.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004444.mm.1 TC1100000571.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0