This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01295920_m1
See other NCEH1 GE Assays ›
Gene Symbol:
Nceh1
Gene Name:
neutral cholesterol ester hydrolase 1
Gene Aliases:
Aadacl1, B230106I24Rik, CPO-BP, Nceh
Chromosome Location:
Chr.3: 27183004 - 27245266 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01295920_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nceh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178772.3 NP_848887.1 3 - 4 514 77
GenBank mRNA AK034339.1 - 3 - 4 441 77
AK045363.1 - 3 - 4 515 77
AK085767.1 - 3 - 4 515 77
AK128968.1 - 3 - 4 525 77
AK135837.1 - 3 - 4 504 77
AK173158.1 - 3 - 4 484 77
BC082569.1 - 3 - 4 497 30550100 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000159.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nceh1

Panther Classification:

Molecular Function -

deacetylase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
lipid metabolic process
xenobiotic metabolic process
metabolic process
lipid catabolic process
SMAD protein signal transduction
phospholipase activity
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
serine hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
phosphate ion binding
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
carboxylic ester hydrolase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300000788.mm.1 NM_178772.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000788.mm.1 NM_178772.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300000788.mm.1 TC0300000159.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300000787.mm.1 NM_178772.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300000787.mm.1 NM_178772.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300000787.mm.1 TC0300000159.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline