This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01295967_g1
See other TMEM9 GE Assays ›
Gene Symbol:
Tmem9
Gene Name:
transmembrane protein 9
Gene Aliases:
1500015G18Rik, AW545782
Chromosome Location:
Chr.1: 136008219 - 136035030 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01295967_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025439.3 NP_079715.1 1 - 2 62 82
GenBank mRNA AK005248.1 - 1 - 2 49 82
AK021225.1 - 1 - 2 49 82
AK154423.1 - 1 - 2 49 82
CA460035.1 - 1 - 2 99 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK005248.1, AK021225.1, AK154423.1, NM_025439.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem9

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100010390.mm.1 NM_001160145.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010390.mm.1 NM_001160145.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010390.mm.1 NM_001160146.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010390.mm.1 NM_001160146.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010390.mm.1 NM_025439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010390.mm.1 NM_025439.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010390.mm.1 TC0100001258.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100010389.mm.1 NM_001160145.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010389.mm.1 NM_001160145.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010389.mm.1 NM_001160146.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010389.mm.1 NM_001160146.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010389.mm.1 NM_025439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010389.mm.1 NM_025439.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010389.mm.1 TC0100001258.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100010385.mm.1 NM_025439.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100010385.mm.1 NM_025439.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100010385.mm.1 TC0100001258.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 433247 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline