This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01295969_m1
See other TMEM9 GE Assays ›
Gene Symbol:
Tmem9
Gene Name:
transmembrane protein 9
Gene Aliases:
1500015G18Rik, AW545782
Chromosome Location:
Chr.1: 136008219 - 136035030 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01295969_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160145.1 NP_001153617.1 4 - 5 504 59
NM_001160146.1 NP_001153618.1 4 - 5 400 59
NM_025439.3 NP_079715.1 4 - 5 369 59
GenBank mRNA AK005248.1 - 4 - 5 356 59
AK021225.1 - 4 - 5 356 59
AK134909.1 - 4 - 5 334 59
AK154423.1 - 4 - 5 356 59
AK218177.1 - 2 - 3 171 59
BC016524.1 - 4 - 5 344 3594475 59
BC020291.1 - 3 - 4 321 59
BC048017.1 - 4 - 5 323 59
BQ918002.1 - 4 - 5 591 59
CA460035.1 - 4 - 5 406 59
CX208138.1 - 4 - 5 400 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem9

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline