This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296179_m1
See other SOAT2 GE Assays ›
Gene Symbol:
Soat2
Gene Name:
sterol O-acyltransferase 2
Gene Aliases:
ACAT2, D15Wsu97e
Chromosome Location:
Chr.15: 102150414 - 102163436 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296179_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Soat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146064.1 NP_666176.1 12 - 13 1301 88
GenBank mRNA AF078751.1 - 12 - 13 1271 88
BC025931.1 - 12 - 13 1301 5099063 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001090.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Soat2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid metabolic process
cholesterol metabolic process
cholesterol efflux
very-low-density lipoprotein particle assembly
cholesterol esterification
cholesterol homeostasis
fatty-acyl-CoA binding
sterol O-acyltransferase activity
O-acyltransferase activity
cholesterol binding
transferase activity
transferase activity, transferring acyl groups
cholesterol O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500008668.mm.1 NM_146064.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008668.mm.1 NM_146064.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008668.mm.1 TC1500001090.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008667.mm.1 NM_146064.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008667.mm.1 NM_146064.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008667.mm.1 TC1500001090.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0