This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296254_m1
See other RARG GE Assays ›
Gene Symbol:
Rarg
Gene Name:
retinoic acid receptor, gamma
Gene Aliases:
Nr1b3, RAR-gamma, RARD, RARgamma2
Chromosome Location:
Chr.15: 102234938 - 102257522 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296254_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rarg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011244.4 NP_035374.3 3 - 4 681 145
GenBank mRNA BC012923.1 - 3 - 4 607 3154037 145
BC013709.1 - 1 - 2 235 3989967 145
M34476.1 - 3 - 4 438 145
X15848.1 - 3 - 4 501 145

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002289.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rarg

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural tube closure
chondrocyte development
glandular epithelial cell development
retinal pigment epithelium development
growth plate cartilage development
growth plate cartilage chondrocyte growth
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
negative regulation of cell proliferation
anterior/posterior pattern specification
regulation of gene expression
positive regulation of gene expression
embryonic camera-type eye development
regulation of myelination
negative regulation of chondrocyte differentiation
response to retinoic acid
embryonic hindlimb morphogenesis
multicellular organism growth
camera-type eye development
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of programmed cell death
steroid hormone mediated signaling pathway
negative regulation of cell differentiation
regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic eye morphogenesis
retinoic acid receptor signaling pathway
reproductive structure development
gland development
retina development in camera-type eye
limb development
face development
bone development
bone morphogenesis
epithelium development
trachea cartilage development
prostate gland epithelium morphogenesis
negative regulation of cartilage development
Harderian gland development
cellular response to retinoic acid
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
retinoic acid receptor activity
protein binding
zinc ion binding
sequence-specific DNA binding
metal ion binding
retinoid X receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0