This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296601_m1
See other TMIGD3 GE Assays ›
Gene Symbol:
Tmigd3
Gene Name:
transmembrane and immunoglobulin domain containing 3
Gene Aliases:
1700001d09rik, 4930578J19Rik
Chromosome Location:
Chr.3: 105870858 - 105924044 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296601_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmigd3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027025.4 NP_081301.2 4 - 5 663 124
GenBank mRNA AK005581.1 - 4 - 5 615 124
BC038878.1 - 3 - 4 543 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001096.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmigd3

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300008858.mm.1 NM_027025.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300008858.mm.1 TC0300001096.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300008859.mm.1 NM_027025.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300008859.mm.1 TC0300001096.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0