This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01296724_m1
See other APPBP2 GE Assays ›
Gene Symbol:
Appbp2
Gene Name:
amyloid beta precursor protein (cytoplasmic tail) binding protein 2
Gene Aliases:
1300003O07Rik, AI465480, PAT1
Chromosome Location:
Chr.11: 85191308 - 85235120 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01296724_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Appbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025825.3 NP_080101.1 1 - 2 455 68
GenBank mRNA AK004888.1 - 1 - 2 466 68
AK005440.1 - 1 - 2 161 68
AK122222.1 - 62 68
AK133853.1 - 1 - 2 447 68
BC018442.1 - 1 - 2 433 3157290 68
BC019152.1 - 1 - 2 456 5065139 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Appbp2

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
intracellular transport
protein binding
microtubule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0