This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01297000_m1
See other ADSL GE Assays ›
Gene Symbol:
Adsl
Gene Name:
adenylosuccinate lyase
Gene Aliases:
Adl, Asl
Chromosome Location:
Chr.15: 80948490 - 80970947 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01297000_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adsl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009634.6 NP_033764.2 7 - 8 844 58
GenBank mRNA AK049372.1 - 7 - 8 816 58
AK051111.1 - 7 - 8 816 58
AK152092.1 - 7 - 8 816 58
AK166019.1 - 7 - 8 812 58
AK168906.1 - 7 - 8 809 58
AK170216.1 - 6 - 7 729 58
AK171725.1 - 7 - 8 825 58
AK190788.1 - 1 - 2 62 58
BC020187.1 - 7 - 8 820 4913637 58
U20225.1 - 7 - 8 820 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000710.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adsl

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide metabolic process
purine nucleotide biosynthetic process
AMP biosynthetic process
response to nutrient
metabolic process
aerobic respiration
purine ribonucleotide biosynthetic process
ribonucleoside monophosphate biosynthetic process
protein tetramerization
catalytic activity
N6-(1,2-dicarboxyethyl)AMP AMP-lyase (fumarate-forming) activity
lyase activity
(S)-2-(5-amino-1-(5-phospho-D-ribosyl)imidazole-4-carboxamido)succinate AMP-lyase (fumarate-forming) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500005393.mm.1 NM_009634.6 Refseq
Affymetrix Mouse Clariom S Array PSR1500005393.mm.1 TC1500000710.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0