This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01297383_g1
See other UBE2A GE Assays ›
Gene Symbol:
Ube2a
Gene Name:
ubiquitin-conjugating enzyme E2A
Gene Aliases:
HHR6A, HR6A, Mhr6a
Chromosome Location:
Chr.X: 36874366 - 36884222 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01297383_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019668.3 NP_062642.1 2 - 3 248 155
GenBank mRNA AF089812.1 - 2 - 3 256 155
AF383148.1 - 2 - 3 256 155
AK133723.1 - 2 - 3 257 155
AK150417.1 - 2 - 3 231 155
AK153327.1 - 2 - 3 231 155
AK160527.1 - 2 - 3 243 155
AK160538.1 - 2 - 3 257 155
AK160545.1 - 2 - 3 255 155
AK164074.1 - 2 - 3 257 155
BC026053.1 - 2 - 3 246 4012075 155

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000329.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2a

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
in utero embryonic development
DNA repair
cellular response to DNA damage stimulus
positive regulation of cell proliferation
response to UV
protein ubiquitination
histone ubiquitination
histone H2A ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein autoubiquitination
maternal process involved in female pregnancy
protein K48-linked ubiquitination
protein K11-linked ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
protein binding
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00002200.mm.1 NM_019668.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00002200.mm.1 TC0X00000329.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0