This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01297483_g1
See other CAR4 GE Assays ›
Gene Symbol:
Car4
Gene Name:
carbonic anhydrase 4
Gene Aliases:
AW456718, Ca4
Chromosome Location:
Chr.11: 84957754 - 84966054 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01297483_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Car4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007607.2 NP_031633.1 6 - 7 691 59
GenBank mRNA AK154754.1 - 6 - 7 657 59
AK165399.1 - 6 - 7 620 59
BC012704.1 - 6 - 7 616 4217743 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001285.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Car4

Gene Ontology Categories:

Function(s) Process(es)

one-carbon metabolic process
regulation of pH
carbon dioxide transport
bicarbonate transport
carbonate dehydratase activity
protein binding
zinc ion binding
lyase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100012164.mm.1 NM_007607.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100012164.mm.1 NM_007607.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100012164.mm.1 TC1100001285.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline