This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01298110_g1
See other ALDOC GE Assays ›
Gene Symbol:
Aldoc
Gene Name:
aldolase C, fructose-bisphosphate
Gene Aliases:
AI847350, AU040929, Aldo3, Scrg2
Chromosome Location:
Chr.11: 78323073 - 78327781 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01298110_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldoc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK005077.1 - 1 - 2 314 68
BC004802.1 - 2 - 3 145 3589325 68
BC008184.1 - 2 - 3 145 3589205 68
S72537.1 - 2 - 3 169 68
X03796.1 - 2 - 3 169 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001156.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldoc

Panther Classification:

Molecular Function -

aldolase

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100010746.mm.1 NM_009657.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100010746.mm.1 TC1100001156.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100010748.mm.1 NM_009657.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100010748.mm.1 TC1100001156.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100010747.mm.1 NM_009657.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100010747.mm.1 TC1100001156.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 429576 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0