This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299345_m1
See other SREK1 GE Assays ›
Gene Symbol:
Srek1
Gene Name:
splicing regulatory glutamine/lysine-rich protein 1
Gene Aliases:
8430401B01, AI450757, AI462342, SRrp508, SRrp86, Sfrs12, Srsf12
Chromosome Location:
Chr.13: 103739348 - 103774603 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299345_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srek1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK039311.1 - 2 - 3 316 94
AK052265.1 - 2 - 3 450 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK039311.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0