This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299376_m1
See other TRAPPC13 GE Assays ›
Gene Symbol:
Trappc13
Gene Name:
trafficking protein particle complex 13
Gene Aliases:
2410002O22Rik, 2610524F24Rik, AI451892, AU017229
Chromosome Location:
Chr.13: 104142153 - 104178703 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299376_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001093759.1 NP_001087228.1 1 - 2 357 113
NM_001093760.1 NP_001087229.1 1 - 2 357 113
NM_025879.2 NP_080155.3 1 - 2 357 113
GenBank mRNA AK010346.1 - 1 - 2 338 113
AK013956.1 - 1 - 2 315 113
AK084373.1 - 1 - 2 307 113
AK163342.1 - 1 - 2 345 113
AK167749.1 - 1 - 2 317 113
BC021756.1 - 1 - 2 298 3584010 113

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK010346.1, AK013956.1, AK084373.1, AK163342.1, AK167749.1, BC021756.1, NM_001093759.1, NM_001093760.1, NM_025879.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc13

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017682.mm.1 NM_001093759.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017682.mm.1 NM_001093759.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017682.mm.1 NM_001093760.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017682.mm.1 NM_001093760.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017682.mm.1 NM_025879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017682.mm.1 NM_025879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017682.mm.1 TC1300002596.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 537276 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0